missing translation for 'onlineSavingsMsg'
Learn More

Guanine, Powder, MP Biomedicals™

Product Code. 11456111 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456111 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456111 Supplier MP Biomedicals Supplier No. 0219069125
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA

Specifications

CAS 73-40-5
Color White
Molecular Formula C5H5N5O
MDL Number MFCD00071533
Quantity 25 g
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
SMILES NC1=NC(=O)C2=C(N1)N=CN2
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
Molecular Weight (g/mol) 151.13
PubChem CID 764
ChEBI CHEBI:16235
Formula Weight 151.13
Grade Highly Purified
Physical Form Powder
Chemical Name or Material Guanine, FTIR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.