missing translation for 'onlineSavingsMsg'
Learn More

Guanine, 98%

Product Code. p-7081853
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11459882

Brand: Thermo Scientific Alfa Aesar A12024.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73-40-5 | C5H5N5O | 151.13 g/mol

Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a nucleobase incorporated into biological nucleic acids. Guanine is suitable reagent used to investigate the mechanism of electrochemical oxidation of guanine and adenine using a glassy carbon microelectrode and cyclic and differential pulse voltammetry. It may be used in the preparation of mixed-sequence peptide nucleic acid (PNA) monomers.

Solubility
Insoluble in water. Soluble in ammonia-water, KOH solutions, dilute acids, and 1 N NaOH (0.1M).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-40-5
Molecular Formula C5H5N5O
Molecular Weight (g/mol) 151.13
MDL Number MFCD00071533
InChI Key UYTPUPDQBNUYGX-UHFFFAOYSA-N
Synonym guanine, 2-amino-6-hydroxypurine, guanin, 2-aminohypoxanthine, mearlmaid, pearl essence, guanine enol, stella polaris, dew pearl, natural pearl essence
PubChem CID 764
ChEBI CHEBI:16235
SMILES NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 9680
Merck Index 14,4564
Solubility Information Insoluble in water. Soluble in ammonia-water,KOH solutions,dilute acids,and 1 N NaOH (0.1M).
IUPAC Name 2-amino-6,7-dihydro-3H-purin-6-one
Formula Weight 151.13
Percent Purity 98%
Chemical Name or Material Guanine
compliance-icons
Product Identifier
  • Guanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Guanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.