missing translation for 'onlineSavingsMsg'
Learn More

Guanine-4,8-13C2,7-15N, TRC

Product Code. 30400118
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400118 25 mg -
30392825 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400118 Supplier Toronto Research Chemicals Supplier No. TRCG83600225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246814-70-9
Molecular Formula C313C2H5N415NO
Molecular Weight (g/mol) 154.1
Synonym 2-Amino-1,9-dihydro-6H-purin-6-one-4,8-13C2,7-15N, 2-Amino-6-hydroxy-1H-purine-4,8-13C2,7-15N, 2-Amino-6-hydroxypurine-4,8-13C2,7-15N, 2-Aminohypoxanthine-4,8-13C2,7-15N, 9H-Guanine-4,8-13C2,7-15N, 2-Amino-hypoxanthine-4,8-13C2,7-15N
IUPAC Name 2-amino-1,9-dihydropurin-6-one
SMILES O=C1NC(N)=N[13C]2=C1[15N]=[13CH]N2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H5N5O/c6-5-9-3-2(4(11)10-5)7-1-8-3/h1H,(H4,6,7,8,9,10,11)/i1+1,3+1,7+1
Formula Weight 154.0532
Chemical Name or Material Guanine-4,8-13C2,7-15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.