missing translation for 'onlineSavingsMsg'
Learn More

Guanidinoacetic-13C Acid, TRC

Product Code. 30399416
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399416 1 mg
30391619 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399416 Supplier Toronto Research Chemicals Supplier No. TRCG8212531MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C C2 H7 N3 O2
Molecular Weight (g/mol) 118.099
Synonym N-(Aminoiminomethyl)-glycine-13C
IUPAC Name 2-carbamimidamidoacetic acid
SMILES NC(=N)N[13CH2]C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C3H7N3O2/c4-3(5)6-1-2(7)8/h1H2,(H,7,8)(H4,4,5,6)/i1+1
Formula Weight 118.057
Chemical Name or Material Guanidinoacetic-13C Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.