missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Thiocyanate, TRC

Product Code. 30011042
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011042 250 mg 250mg
30003789 2.5 g 2.5g
30006947 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011042 Supplier Toronto Research Chemicals Supplier No. TRCG838723250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-84-0
Molecular Formula C H5 N3 . C H N S
Molecular Weight (g/mol) 118.16
Synonym Guanidine Thiocyanate, Thiocyanic acid, compd. with guanidine (1:1), Guanidine thiocyanate (6CI), Guanidine, monothiocyanate (8CI,9CI), Buffer AVL, Buffer ML, Buffer MTL, Buffer RLT, GTC, Guanidine isothiocyanate, Guanidinium isothiocyanate, Guanidinium thiocyanate, NSC 2119, RLT
IUPAC Name guanidine;thiocyanic acid
SMILES NC(=N)N.SC#N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H
Formula Weight 118.0313
Chemical Name or Material Guanidine Thiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.