Learn More
Thermo Scientific Chemicals Guanidine hydrochloride, ultrapure, 99%
£48.90 - £277.00
Chemical Identifiers
CAS | 50-01-1 |
---|---|
Molecular Formula | CH6ClN3 |
Molecular Weight (g/mol) | 95.53 |
MDL Number | MFCD00013026 |
InChI Key | PJJJBBJSCAKJQF-UHFFFAOYSA-N |
Synonym | guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride |
PubChem CID | 5742 |
ChEBI | CHEBI:32735 |
IUPAC Name | guanidine;hydrochloride |
SMILES | C(=N)(N)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436929
|
Thermo Scientific Chemicals
J65485.22 |
100 g |
£48.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446929
|
Thermo Scientific Chemicals
J65485.36 |
500 g |
£164.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15456929
|
Thermo Scientific Chemicals
J65485.A1 |
1 kg |
£277.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Guanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGuanidine hydrochloride is used for purification of proteins and nucleic acids. It can be used as a medicine, organic synthesis intermediate and is used in dyes. It acts as an intermediate in the preparation of sulfadiazine, which is an important raw material for sulfamethyldiazine, sulfamethazine and folic acid. It is utilized to synthesis of 2-amino-pyrimidine, 2- amino -6- methyl-pyrimidine and 2-amino -4,6 - dimethyl-pyrimidine. Also used in RNA isolation to dissociate nucleoproteins and inhibit RNase.
Solubility
Soluble in water and ethanol.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.May agglomerate upon storage.
Chemical Identifiers
50-01-1 | |
95.53 | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
5742 | |
guanidine;hydrochloride |
CH6ClN3 | |
MFCD00013026 | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
CHEBI:32735 | |
C(=N)(N)N.Cl |
Specifications
50-01-1 | |
White | |
CH6ClN3 | |
3591990 | |
14,4562 | |
Soluble in water and ethanol. | |
C(=N)(N)N.Cl | |
95.53 | |
CHEBI:32735 | |
99% | |
Guanidine hydrochloride |
185°C to 188°C | |
1.344 | |
MFCD00013026 | |
Hygroscopic | |
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
guanidine;hydrochloride | |
5742 | |
95.53 | |
Powder | |
Ultrapure |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H500
missing translation for 'einecsNumber' : 200-002-3
missing translation for 'rtecsNumber' : MF4300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only