missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride Optical Density Grade MP Biomedicals

Product Code. 11473731 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473731 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473731 Supplier MP Biomedicals Supplier No. 0215121080
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50-01-1
Density 1.345g/cm3 at 20 °C
Molecular Formula CH6ClN3
Quantity 100 g
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
SMILES C(=N)(N)N.Cl
IUPAC Name guanidine;hydrochloride
Molecular Weight (g/mol) 95.53
PubChem CID 5742
ChEBI CHEBI:32735
Formula Weight 95.5
Grade Optical Density
Chemical Name or Material Guanidine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.