missing translation for 'onlineSavingsMsg'
Learn More

Guanidine hydrochloride, MedChemExpress

Product Code. 16428388
Encompass_Preferred
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 16428388

missing translation for 'mfr': Medchem Express HYB0178A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Guanidine hydrochloride (Guanidinium chloride) a strong chaotrope, is also a strong denaturant of proteins.

Guanidine hydrochloride (Guanidinium chloride) a strong chaotrope, is also a strong denaturant of proteins.

Specifications

Chemical Name or Material Guanidine hydrochloride
Physical Form Solid
CAS 50-01-1
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula CH6ClN3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Guanidinium chlorideAminoformamidine hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (1046.79 mM) ∣H2O : ≥ 50 mg/mL (523.40 mM)
SMILES NC(N)=N.Cl
Molecular Weight (g/mol) 95.53
Formula Weight 95.53
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.