missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
Guanidine hydrochloride (Guanidinium chloride) a strong chaotrope, is also a strong denaturant of proteins.
Specifications
Specifications
| Chemical Name or Material | Guanidine hydrochloride |
| Physical Form | Solid |
| CAS | 50-01-1 |
| Color | White |
| Quantity | 50 g |
| Purity Grade Notes | Research |
| Molecular Formula | CH6ClN3 |
| For Use With (Application) | Metabolism-protein/nucleotide metabolism |
| Synonym | Guanidinium chlorideAminoformamidine hydrochloride |
| Solubility Information | DMSO : ≥ 100 mg/mL (1046.79 mM) ∣H2O : ≥ 50 mg/mL (523.40 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction