missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, 99+%, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
3 kg
Unit Size:
100g
1kg
25g
3kg
500g
Specifications
Specifications
| Assay Percent Range | 99+% |
| Quantity | 25 g |
| Formula Weight | 95.53 |
| Chemical Name or Material | Guanidine Hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction