missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, >99.5%, Molecular Biology Grade

Product Code. 15835228 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Poly pail
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15835228 100 g Plastic bottle 100g
15845228 500 g Plastic bottle 500g
15815368 1 kg Plastic bottle 1kg
15825368 5 kg Poly pail 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15835228 Supplier Thermo Scientific Supplier No. J75823.22
  • ONLINE EXCLUSIVE PRICE
    £53.00
    £47.65 / 100g
    Save £5.35
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol

  • Used for extraction of RNA.
  • Preparation of high molecular weight genomic DNA.
  • Solubilizing inclusion bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used for extraction of RNA
  • Preparation of high molecular weight genomic DNA
  • Solubilizing inclusion bodies
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Melting Point 185°C to 188°C
Density 1.344
Packaging Plastic bottle
Linear Formula (NH2)2C=NH·HCl
Quantity 100 g
Beilstein 3591990
Formula Weight 95.53
Percent Purity ≥99.5%
Grade Molecular Biology
Physical Form Crystals
Chemical Name or Material Guanidine Hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.