missing translation for 'onlineSavingsMsg'
Learn More

Guanidine-d5 DCl, CDN

Product Code. 30324703
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324703 1 g 1g
30315312 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324703 Supplier CDN Supplier No. CDND11121G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanidine-d5 Hydrochloride-d
CAS 108694-93-5
Molecular Formula C D5 N3 . Cl D
Quantity 1 g
Synonym Hydrochloric acid-d, compd. with guanidine-d5 (1:1) (9 CI), Guanidine-d5, mono(hydrochloride-d) (9 CI), Guanidinium chloride-d6, Guanidine-D5 Deuterochloride
InChI Formula InChI=1 S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1 H/i/hD6
SMILES [2 H]Cl.[2 H]N=C(N([2 H])[2 H])N([2 H])[2 H]
IUPAC Name (2 H)chlorane;1,1,2,3,3-pentadeuterioguanidine
Molecular Weight (g/mol) 101.568
Formula Weight 101.063 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.