missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Carbonate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Guanidine Carbonate |
| CAS | 593-85-1 |
| Molecular Formula | 2(CH5N3) . (CH2O3) |
| Quantity | 25 g |
| Synonym | Bisguanidinium Carbonate, Diguanidinium Carbonate, Guanidinium Carbonate |
| InChI Formula | InChI=1S/2CH5N3.CH2O3/c3*2-1(3)4/h2*(H5,2,3,4);(H2,2,3,4) |
| SMILES | NC(=N)N.NC(=N)N.OC(=O)O |
| IUPAC Name | carbonic acid;guanidine |
| Molecular Weight (g/mol) | 180.17 |
| Formula Weight | 180.0971 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction