missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanfacine hydrochloride, MedChemExpress
Description
Guanfacine hydrochloride, an anti-hypertensive agent, is a selective α2A-adrenoceptor agonist with Kd of 31 nM and displays 60-fold selectivity over α2B-adrenoceptors.
Specifications
Specifications
| Chemical Name or Material | Guanfacine hydrochloride |
| Physical Form | Solid |
| CAS | 29110-48-3 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H10Cl3N3O |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 30 mg/mL (106.18 mM) ∣H2O : 20 mg/mL (70.78 mM; Need ultrasonic) |
| SMILES | O=C(NC(N)=N)CC1=C(Cl)C=CC=C1Cl.[H]Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?