missing translation for 'onlineSavingsMsg'
Learn More

Guancydine, MedChemExpress

Product Code. 16466137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466137 5 mg 5mg
16403418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466137 Supplier Medchem Express Supplier No. HY101554

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guancydine (Guancidine) is an antihypertensive agent.

Guancydine (Guancidine) is an antihypertensive agent.

Specifications

Chemical Name or Material Guancydine
Physical Form Solid
CAS 1113-10-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C7H14N4
For Use With (Application) Neuroscience-Neuromodulation
Synonym Guancidine
Solubility Information DMSO : ≥ 125 mg/mL (810.58 mM)
SMILES N=C(NC(C)(C)CC)NC#N
Molecular Weight (g/mol) 154.21
Formula Weight 154.21
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.