missing translation for 'onlineSavingsMsg'
Learn More

Guanabenz acetate, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18752574
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18752574

Brand: Tocris Bioscience 0885/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

α2 agonist. Also I2 selective ligand

α2-adrenergic agonist and IGRS (imidazoline I2 binding site) selective ligand.
TRUSTED_SUSTAINABILITY

Specifications

CAS 23256-50-0
Molecular Formula C10H12Cl2N4O2
Quantity 100 mg
Synonym guanabenz wy-8678 acetate, 2,6-dichlorobenzylidene-amino guanidine, acetic acid; 2-2,6-dichlorophenyl methylideneamino guanidine, n-2,6-dichlorophenyl methylidene amino guanidine; acetic acid
InChI Key MCSPBPXATWBACD-UHFFFAOYSA-N
SMILES CC(=O)O.C1=CC(=C(C(=C1)Cl)C=NN=C(N)N)Cl
IUPAC Name acetic acid;2-[(2,6-dichlorophenyl)methylideneamino]guanidine
Molecular Weight (g/mol) 291.132
PubChem CID 6852405
Formula Weight 291.14
Percent Purity >98%
Chemical Name or Material Guanabenz acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.