missing translation for 'onlineSavingsMsg'
Learn More

Guaifenesin, Mikromol™

Product Code. 17930486
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930486 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17930486 Supplier Mikromol Supplier No. MM0354.00
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Guaifenesin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guaifenesin
CAS 93-14-1
Analyte or Component Names Guaifenesin
Shipping Condition Room Temperature
Molecular Formula C10 H14 O4
Certifications/Compliance ISO 17025
Synonym 1,2-Dihydroxy-3-(2-methoxyphenoxy)propane, 2-G, 3-(2-Methoxyphenoxy)-1,2-propanediol, 3-(o-Methoxyphenoxy)-1,2-propanediol, Actifed C, Aeronesin, Amonidren, Aresol, Calmipan, Colrex Expectorant, Creson, Dilyn, Equicol, Glycerin guaiacolate, Glycerol guaiacolate, Glyceryl guaiacol ether, Glyceryl guaiacolate, Glyceryl guaiacolate ether, Glyceryl guaiacyl ether, Glycerylguaiacol, Glycodex, Glycotuss, Guaiacol glycerin ether, Guaiacol glycerol ether, Guaiacol glyceryl ether, Guaiacuran, Guaiacurane, Guaiacyl glyceryl ether, Guaiamar, Guaianesin, Guaifenesin, Guaifenesine, Guaiphenesin, Guaiphenesine, Guajacuran, Guanar, Guayanesin, Hustosil, Hytuss, Methoxypropanediol, Methphenoxydiol, Miocurin, Mucinex, Muskurelax, My 301, Myocain, Myocaine, Myoscain, NSC 62112, Neuroton, Neurotone, Oresol, Oreson, Organidin NR, Relaxil G, Reorganin, Respenyl, Resyl, Ritussin, Robitussin, SL 90, Sirotol, Tenntus, Tenntuss, Trecid, Tulyn, XL 90, o-Methoxyphenyl glyceryl ether, racemic-Guaifenesin, 1,2-Propanediol, 3-(o-methoxyphenoxy)- (6CI,8CI), Giafen, Glycerol alpha-(2-methoxyphenyl) ether, Glycerol alpha-(o-methoxyphenyl)ether, Glycerol alpha-guaiacyl ether, alpha-Glyceryl guaiacol ether, alpha-Glyceryl guaiacolate ether
InChI Formula InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12H,6-7H2,1H3
SMILES COc1ccccc1OCC(O)CO
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
Molecular Weight (g/mol) 198.22
Quantity 500 mg
Formula Weight 198.0892 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.