missing translation for 'onlineSavingsMsg'
Learn More

Guaifenesin, MedChemExpress

Product Code. p-200099137
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490848 5 g 5g
16412119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490848 Supplier Medchem Express Supplier No. HYB0264

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Guaifenesin (Guaiacol glyceryl ether), a constituent of guaiac resin from the wood of Guajacum officinale Linné, is an expectorant. Guaifenesin can alleviate cough discomfortby increasing sputum volume and decreasing its viscosity, thereby promoting effective cough.

Guaifenesin (Guaiacol glyceryl ether), a constituent of guaiac resin from the wood of Guajacum officinale Linné, is an expectorant. Guaifenesin can alleviate cough discomfortby increasing sputum volume and decreasing its viscosity, thereby promoting effective cough.

Specifications

Chemical Name or Material Guaifenesin
CAS 93-14-1
Color White
Purity Grade Notes Research
Molecular Formula C10H14O4
Quantity 5 g
Synonym Guaiacol glyceryl ether Guaiphenesin Glycerol guaiacolate
Solubility Information DMSO : 100 mg/mL (504.49 mM; Need ultrasonic and warming)
SMILES OCC(O)COC1=CC=CC=C1OC
Molecular Weight (g/mol) 198.22
Formula Weight 198.22
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.