missing translation for 'onlineSavingsMsg'
Learn More

Guaifenesin, LoGiCal

Product Code. 30460007
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460007 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460007 Supplier LoGiCal Supplier No. LGCFOR0354.00
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guaifenesin
CAS 93-14-1
Analyte or Component Names Guaifenesin
Shipping Condition Room Temperature
Molecular Formula C10 H14 O4
Certifications/Compliance ISO 9001
Synonym 1,2-Dihydroxy-3-(2-methoxyphenoxy)propane, 2-G, 3-(2-Methoxyphenoxy)-1,2-propanediol, 3-(o-Methoxyphenoxy)-1,2-propanediol, Actifed C, Aeronesin, Amonidren, Aresol, Calmipan, Colrex Expectorant, Creson, Dilyn, Equicol, Glycerin guaiacolate, Glycerol guaiacolate, Glyceryl guaiacol ether, Glyceryl guaiacolate, Glyceryl guaiacolate ether, Glyceryl guaiacyl ether, Glycerylguaiacol, Glycodex, Glycotuss, Guaiacol glycerin ether, Guaiacol glycerol ether, Guaiacol glyceryl ether, Guaiacuran, Guaiacurane, Guaiacyl glyceryl ether, Guaiamar, Guaianesin, Guaifenesin, Guaifenesine, Guaiphenesin, Guaiphenesine, Guajacuran, Guanar, Guayanesin, Hustosil, Hytuss, Methoxypropanediol, Methphenoxydiol, Miocurin, Mucinex, Muskurelax, My 301, Myocain, Myocaine, Myoscain, NSC 62112, Neuroton, Neurotone, Oresol, Oreson, Organidin NR, Relaxil G, Reorganin, Respenyl, Resyl, Ritussin, Robitussin, SL 90, Sirotol, Tenntus, Tenntuss, Trecid, Tulyn, XL 90, o-Methoxyphenyl glyceryl ether, racemic-Guaifenesin, 1,2-Propanediol, 3-(o-methoxyphenoxy)- (6CI,8CI), Giafen, Glycerol alpha-(2-methoxyphenyl) ether, Glycerol alpha-(o-methoxyphenyl)ether, Glycerol alpha-guaiacyl ether, alpha-Glyceryl guaiacol ether, alpha-Glyceryl guaiacolate ether
InChI Formula InChI=1S/C10H14O4/c1-13-9-4-2-3-5-10(9)14-7-8(12)6-11/h2-5,8,11-12H,6-7H2,1H3
Solution Type Neat
SMILES COc1ccccc1OCC(O)CO
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
Industry Type Clinical Testing
Molecular Weight (g/mol) 198.22
Quantity 10 mg
Formula Weight 198.0892 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.