missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol, SLR, Fisher Chemical

Product Code. 10726101 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10726101 100 g Amber glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10726101 Supplier Fisher Chemical Supplier No. G/0950/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 90-05-1
Melting Point 27°C
Boiling Point 205°C
Packaging Amber glass bottle
Molecular Formula C7H8O2
MDL Number 2185
Quantity 100 g
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1O
IUPAC Name 2-methoxyphenol
Molecular Weight (g/mol) 124.139
PubChem CID 460
ChEBI CHEBI:28591
Formula Weight 124.14g/mol
Physical Form Low Melting Solid
Show More Show Less
compliance-icons
Product Identifier
  • Guaiacol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.