missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol MP Biomedicals

Product Code. 11433772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433772 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433772 Supplier MP Biomedicals Supplier No. 0215727790
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 90-05-1
Density 4.27 (air = 1)
Molecular Formula C7H8O2
Quantity 500 g
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1O
IUPAC Name 2-methoxyphenol
Molecular Weight (g/mol) 124.139
PubChem CID 460
ChEBI CHEBI:28591
Formula Weight 124.15
Chemical Name or Material Guaiacol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.