missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol-Beta-D-glucopyranoside, TRC

Product Code. 30394728
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394728 50 mg
30389647 5 mg
30398225 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394728 Supplier Toronto Research Chemicals Supplier No. TRCG24556550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6092-24-6
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
Synonym o-Methoxyphenyl beta-D-glucopyranoside, 2-Methoxyphenyl beta-D-glucopyranoside
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(2-methoxyphenoxy)oxane-3,4,5-triol
SMILES COc1ccccc1O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H18O7/c1-18-7-4-2-3-5-8(7)19-13-12(17)11(16)10(15)9(6-14)20-13/h2-5,9-17H,6H2,1H3/t9-,10-,11+,12-,13-/m1/s1
Formula Weight 286.1053
Chemical Name or Material Guaiacol-beta-D-glucopyranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.