Learn More
Guaiacol, 99+%, Thermo Scientific Chemicals
£25.50 - £214.00
Chemical Identifiers
CAS | 90-05-1 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.14 |
InChI Key | LHGVFZTZFXWLCP-UHFFFAOYSA-N |
Synonym | guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid |
PubChem CID | 460 |
ChEBI | CHEBI:28591 |
IUPAC Name | 2-methoxyphenol |
SMILES | COC1=CC=CC=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316610
|
Thermo Scientific Acros
120192500 |
250 g | Glass bottle |
£25.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10744991
|
Thermo Scientific Acros
120190010 |
1 kg | Glass bottle |
£62.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944751
|
Thermo Scientific Acros
120190050 |
5 kg | Plastic drum |
£214.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90-05-1 | |
124.14 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
CHEBI:28591 | |
COC1=CC=CC=C1O |
C7H8O2 | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
460 | |
2-methoxyphenol |
Specifications
90-05-1 | |
Colorless to Yellow | |
205.0°C | |
Authentic | |
C7H8O2 | |
1.5430 to 1.5450 | |
15,4589 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
2-methoxyphenol | |
460 | |
124.14 | |
Liquid After Melting |
27.0°C to 29.0°C | |
1.1200g/mL | |
82°C | |
99% min. (GC) | |
2-(CH3O)C6H4OH | |
06,768 | |
1.12 | |
Solubility in water: 17g/L (15°C). Other solubilities: miscible with glacial acetic acid, miscible with alcohol, chloroform and ether, slightly soluble in petroleum ether | |
COC1=CC=CC=C1O | |
124.14 | |
CHEBI:28591 | |
99+% | |
Guaiacol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-964-7
- Guaiacol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only