missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol, 99.54%, MP Biomedicals™

Product Code. 11423772 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
This item is not returnable. View return policy

Product Code. 11423772

Brand: MP Biomedicals 0215727783

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Color Colorless to Yellow
Density 1.129g/cm3 at 21°C
Assay Percent Range 0.9954
Quantity 250 g
Formula Weight 124.1
Physical Form Crystalline Solid to Liquid
Chemical Name or Material Guaiacol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.