missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol, 99.54%, MP Biomedicals™

Product Code. 11413772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413772 100 g 100g
11423772 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413772 Supplier MP Biomedicals Supplier No. 0215727780
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Color Colorless to Yellow
Density 1.129g/cm3 at 21°C
Assay Percent Range 0.9954
Quantity 100 g
Formula Weight 124.1
Physical Form Crystalline Solid to Liquid
Chemical Name or Material Guaiacol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.