missing translation for 'onlineSavingsMsg'
Learn More

Guacetisal, MedChemExpress

Product Code. 16497358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497358 10 mg 10mg
16497326 100 mg 100mg
16463576 25 mg 25mg
16463387 50 mg 50mg
16409847 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497358 Supplier Medchem Express Supplier No. HY17477
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Guacetisal is obtained from the esterification of acetylsalicylic acid with guaiacol which has the potential for chronic bronchitis treatment extracted from patent CN 106866420 A.

Guacetisal is obtained from the esterification of acetylsalicylic acid with guaiacol which has the potential for chronic bronchitis treatment extracted from patent CN 106866420 A.

Specifications

Chemical Name or Material Guacetisal
CAS 55482-89-8
Color White
Purity Grade Notes Research
Molecular Formula C16H14O5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (349.31 mM; Need ultrasonic)
SMILES O=C(OC1=CC=CC=C1OC)C2=CC=CC=C2OC(C)=O
Molecular Weight (g/mol) 286.28
Formula Weight 286.28
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.