missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTSE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526879
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
The protein encoded by this gene is only expressed in the S and G2 phases of the cell cycle, where it colocalizes with cytoplasmic tubulin and microtubules. In response to DNA damage, the encoded protein accumulates in the nucleus and binds the tumor suppressor protein p53, shuttling it out of the nucleus and repressing its ability to induce apoptosis.
Specifications
| GTSE1 | |
| Polyclonal | |
| Unconjugated | |
| GTSE1 | |
| B99; G two S phase expressed protein 1; G2 and S phase-expressed protein 1; G-2 and S-phase expressed 1; G2 and S-phase expressed 1; GTSE1; GTSE-1; Protein B99; Protein B99 homolog; RGD1563164 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 51512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NYZ3 | |
| GTSE1 | |
| KLH conjugated synthetic peptide between 690-717 amino acids from the C-terminal region of human GTSE1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction