missing translation for 'onlineSavingsMsg'
Learn More

GTS-21 dihydrochloride, MedChemExpress

Product Code. 16446496
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446496

Brand: Medchem Express HY14564A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GTS-21 dihydrochloride is a selective alpha7 nicotinic acetylcholine receptor (α7-nAChR) agonist with anti‐inflammatory and cognition‐enhancing activities. GTS-21 dihydrochloride is also a α4β2 (Ki=20 nM for humanα4β2) and 5-HT3A receptor (IC50=3.1 μM) antagonist.

GTS-21 dihydrochloride is a selective alpha7 nicotinic acetylcholine receptor (α7-nAChR) agonist with anti‐inflammatory and cognition‐enhancing activities. GTS-21 dihydrochloride is also a α4β2 (Ki=20 nM for humanα4β2) and 5-HT3A receptor (IC50=3.1 μM) antagonist.

Specifications

Chemical Name or Material GTS-21 dihydrochloride
CAS 156223-05-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H22Cl2N2O2
Quantity 100 mg
Synonym DMXB-A DMBX-anabaseine
Solubility Information H2O : 50 mg/mL (131.13 mM; Need ultrasonic) ∣DMSO : 16.5 mg/mL (43.27 mM; Need ultrasonic and warming)
SMILES COC1=CC(OC)=CC=C1/C=C2CCCN=C\2C3=CC=CN=C3.Cl[H].Cl[H]
Molecular Weight (g/mol) 381.3
Formula Weight 381.3
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.