missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57545
This item is not returnable.
View return policy
Description
GTPBP9 Polyclonal specifically detects GTPBP9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GTPBP9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 100%; Human: 100%; Chicken: 100%; Sumatran orangutan: 100%; Bovine: 100%; Mouse: 100%; Endeis laevis: 92%; Nicoletia meinerti: 92%; Skogsbergia lerneri: 92%; Streptocephalus seali: 92%; Brucella sp. 83/13: 92%; Brucella pinnipedialis M292/94/1: 92%; Brucella melitensis bv. 1 str. Rev.1: 92%; Brucella melitensis bv. 3 str. Ether: 92%; Woodlice: 92%; Brine shrimp: 92%; Ammothea hilgendorfi: 92%; Brucella abortus NCTC 8038: 92%; Brucella melitensis bv. 1 str. 16M: 92%; Brucella suis bv. 4 str. 40: 92%; Brucella melitensis bv. 2 str. 63/9: 92%; Brucella suis bv. 5 str. 513: 92%; Brucella suis bv. 3 str. 686: 92%; Brucella sp. F5/99: 92%; Achelia echinata: 92%; Brucella ceti M490/95/1: 92%; Gluconacetobacter hansenii ATCC 23769: 92%; Brucella sp. BO2: 92%; Sphingomonas sp. SKA58: 92%; Brucella abortus: 92%; Gluconobacter suboxydans: 92%; Zymomonas mobilis: 92%; Brucella suis: 92%; Brucella melitensis: 92%; Sinorhizobium meliloti: 92%; Harpegnathos saltator: 92%; Ahrensia sp. R2A130: 92%; Brucella sp. NVSL 07-0026: 92%; Red flour beetle: 92%; Brucella abortus bv. 5 str. B3196: 92%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9NTK5 | |
| OLA1 | |
| Synthetic peptides corresponding to GTPBP9 (GTP-binding protein 9 (putative)) The peptide sequence was selected from the N terminal of GTPBP9. Peptide sequence QGLGNAFLSHISACDGIFHLTRAFEDDDITHVEGSVDPIRDIEIIHEELQ. | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 29789 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction