missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20133-1-AP-150UL
This item is not returnable.
View return policy
Description
Small G proteins, such as GTPBP5, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 ).Specifications
| GTPBP5 | |
| Polyclonal | |
| Unconjugated | |
| MTG2 | |
| GTP binding protein 5, GTPBP5, ObgH1, Protein obg homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26164, 296462, 52856 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H4K7 | |
| Mtg2 | |
| GTPBP5 Fusion Protein Ag14015 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction