missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTPBP4 Recombinant Rabbit Monoclonal Antibody (JE47-47)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534853
This item is not returnable.
View return policy
Description
Positive Control: 293T cell, human liver cancer tissue, human thyroid gland tissue, human skin tissue, human breast tissue, thp-1.
GTP-binding proteins are GTPases and function as molecular switches that can flip between two states: active, when GTP is bound, and inactive, when GDP is bound. 'Active' in this context usually means that the molecule acts as a signal to trigger other events in the cell. When an extracellular ligand binds to a G-protein-linked receptor, the receptor changes its conformation and switches on the trimeric G proteins that associate with it by causing them to eject their GDP and replace it with GTP. The switch is turned off when the G protein hydrolyzes its own bound GTP, converting it back to GDP. But before that occurs, the active protein has an opportunity to diffuse away from the receptor and deliver its message for a prolonged period to its downstream target.
Specifications
| GTPBP4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9BZE4 | |
| Gtpbp4 | |
| Recombinant protein within Human GTPBP4 aa 1-131. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE47-47 | |
| Unconjugated | |
| Gtpbp4 | |
| 2610028C09Rik; chronic renal failure gene protein; chronic renal failure protein; Crfg; fi28d07; FLJ10686; FLJ10690; FLJ39774; G protein-binding protein CRFG; GTP binding protein 3; GTP binding protein 4; GTP binding protein NGB; GTP-binding protein NGB; Gtpbp3; Gtpbp4; NGB; Nog1; nucleolar GTP-binding protein 1; Nucleolar GTP-binding protein 1 (Chronic renal failure gene protein) (GTP-binding protein NGB); Unknown (protein for IMAGE:7956347); wu:fi28d07; zgc:55757 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23560 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction