missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTPBP4 Recombinant Rabbit Monoclonal Antibody (23GB6455), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GTPBP4 |
|---|---|
| Clone | 23GB6455 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087847
![]() |
Invitrogen™
MA553054 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088146
|
Invitrogen™
MA553055 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTPBP4 Recombinant Monoclonal antibody specifically detects GTPBP4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GTPBP4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q99ME9, Q99P77, Q9BZE4 | |
| 114300, 23560, 69237 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6455 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2610028C09Rik; chronic renal failure gene protein; chronic renal failure protein; Crfg; fi28d07; FLJ10686; FLJ10690; FLJ39774; G protein-binding protein CRFG; GTP binding protein 3; GTP binding protein 4; GTP binding protein NGB; GTP-binding protein NGB; Gtpbp3; Gtpbp4; NGB; Nog1; nucleolar GTP-binding protein 1; Nucleolar GTP-binding protein 1 (Chronic renal failure gene protein) (GTP-binding protein NGB); Unknown (protein for IMAGE:7956347); wu:fi28d07; zgc:55757 | |
| A synthesized peptide derived from human GTPBP4 (50-100AA). | |
| Primary | |
| -20°C | |
| Gtpbp4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title