missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTPBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531323
This item is not returnable.
View return policy
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Predicted reactivity: Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This locus encodes a GTP-binding protein. The encoded protein is localized to the mitochondria and may play a role in mitochondrial tRNA modification. Polymorphisms at this locus may be associated with severity of aminoglycoside-induced deafness, a disease associated with a mutation in the 12S rRNA. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| GTPBP3 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP3 | |
| 2410009F13Rik; AI607903; COXPD23; GTP binding protein 3; GTP binding protein 3 (mitochondrial); GTPBG3; GTP-binding protein 3; Gtpbp3; Mitochondrial GTP-binding protein 1; MSS1; MTGP1; THDF1; tRNA modification GTPase GTPBP3, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84705 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q969Y2 | |
| GTPBP3 | |
| Recombinant protein encompassing a sequence within the center region of human GTPBP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction