missing translation for 'onlineSavingsMsg'
Learn More

GTP binding protein era homolog Antibody, Novus Biologicals™

Product Code. 18357489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357489 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357489 Supplier Novus Biologicals Supplier No. H00026284B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GTP binding protein era homolog Polyclonal antibody specifically detects GTP binding protein era homolog in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTP binding protein era homolog
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005693.1
Gene Alias CEGA, Conserved ERA-like GTPase, ERA, Era (E. coli G-protein homolog)-like 1, Era G-protein-like 1 (E. coli), ERAL1A, ERA-like protein 1, ERA-W, GTPase Era, mitochondrial, GTPase, human homolog of E. coli essential cell cycle protein Era, GTP-binding protein era homolog, HERA, H-ERA, HERA-A, HERA-B
Host Species Mouse
Immunogen ERAL1 (NP_005693.1, 1 a.a. - 437 a.a.) full-length human protein. MAAPSWRGARLVQSVLRVWQVGPHVARERVIPFSSLLGFQRRCVSCVAGSAFSGPRLASASRSNGQGSALDHFLGFSQPDSSVTPCVPAVSMNRDEQDVLLVHHPDMPENSRVLRVVLLGAPNAGKSTLSNQLLGRKVFPVSRKVHTTRCQALGVITEKETQVILLDTPGIISPGKQKRHHLELSLLEDPWKSMESADLVVVLVDVSDKWTRNQLSPQLLRCLTKYSQIPSVLVMNKVDCLKQKSVLLELTAALTEGVVNGKKLKMRQAFHSHPGTHCPSPAVKDPNTQSVGNPQRIGWPHFKEIFMLSALSQEDVKTLKQYLLTQAQPGPWEYHSAVLTSQTPEEICANIIREKLLEHLPQEVPYNVQQKTAVWEEGPGGELVIQQKLLVPKESYVKLLIGPKGHVISQIAQEAGHDLMDIFLCDVDIRLSVKLLK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26284
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.