missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17653-1-AP-150UL
This item is not returnable.
View return policy
Description
The Integrator complex contains at least 12 subunits and associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates the 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690). INTS11, or CPSF3L, is the catalytic subunit of the Integrator complex.Specifications
| GTF3C4 | |
| Polyclonal | |
| Unconjugated | |
| GTF3C4 | |
| GTF3C4, KAT12, TF3C delta, TFIII90, TFIIIC 90 kDa subunit, TFiiiC2 90, TFIIIC90, TFIIICdelta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 269252, 685539, 9329 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BMQ2, Q9UKN8 | |
| GTF3C4 | |
| GTF3C4 Fusion Protein Ag11898 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction