missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | GTF3C2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18479220
|
Novus Biologicals
NBP1-83062-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18216508
|
Novus Biologicals
NBP1-83062 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF3C2 Polyclonal specifically detects GTF3C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GTF3C2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 2976 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LRREPMLRMQEGEGHSQLCLDRLQLEAIHKVRFSPNLDSYGWLVSGGQSGLVRIHFVRGLASPLGHRMQLESRAHFNAMFQPSSPTRRPGFSPTSH | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| general transcription factor 3C polypeptide 2, general transcription factor IIIC, polypeptide 2, beta 110kDa, KIAA0011general transcription factor IIIC, polypeptide 2 (beta subunit, 110kD), TF3C-beta, TFIIIC 110 kDa subunit, TFIIIC110Transcription factor IIIC 110 kDa subunit, TFIIIC-BETA, Transcription factor IIIC subunit beta | |
| GTF3C2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title