missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106758
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GTF3A.
The product of this gene is a zinc finger protein with nine Cis[2]-His[2] zinc finger domains. It functions as an RNA polymerase III transcription factor to induce transcription of the 5S rRNA genes. The protein binds to a 50 bp internal promoter in the 5S genes called the internal control region (ICR), and nucleates formation of a stable preinitiation complex. This complex recruits the TFIIIC and TFIIIB transcription factors and RNA polymerase III to form the complete transcription complex. The protein is thought to be translated using a non-AUG translation initiation site in mammals based on sequence analysis, protein homology, and the size of the purified protein.
Specifications
| GTF3A | |
| Polyclonal | |
| Unconjugated | |
| GTF3A | |
| AP2; general transcription factor IIIA; GTF3A; TF3A; TFIIIA; Transcription factor IIIA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2971 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92664 | |
| GTF3A | |
| A synthesized peptide derived from human GTF3A(Accession Q92664), corresponding to amino acid residues V6-K56. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction