missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GTF3A Polyclonal antibody specifically detects GTF3A in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | GTF3A |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | general transcription factor IIIA, TFIIIAAP2, transcription factor IIIA |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GTF3A (NP_002088.2).,, Sequence:, MDPPAVVAESVSSLTIADAFIAAGESSAPTPPRPALPRRFICSFPDCSANYSKAWKLDAHLCKHTGERPFVCDYEGCGKAFIRDYHLSRHILTHTGEKPF |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?