missing translation for 'onlineSavingsMsg'
Learn More

GTF2IRD2 Mouse anti-Human, Clone: PCRP-GTF2IRD2-1B4, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07400-20UG

 View more versions of this product

Product Code. 18301162

  • £196.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GTF2IRD2 Monoclonal specifically detects GTF2IRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GTF2IRD2
Monoclonal
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha
Recombinant full-length human GTF2IRD2 protein
20 μg
Primary
Human
Purified
Western Blot, Flow Cytometry, Peptide Array
PCRP-GTF2IRD2-1B4
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
84163
Store at 4°C.
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.