missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2IRD2 Mouse anti-Human, Clone: PCRP-GTF2IRD2-1B4, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | GTF2IRD2 |
|---|---|
| Clone | PCRP-GTF2IRD2-1B4 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
Description
GTF2IRD2 Monoclonal specifically detects GTF2IRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.Specifications
| GTF2IRD2 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha | |
| Recombinant full-length human GTF2IRD2 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-GTF2IRD2-1B4 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 84163 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title