missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2IRD2 Antibody (PCRP-GTF2IRD2-1B12), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08512MFV610
This item is not returnable.
View return policy
Description
GTF2IRD2 Monoclonal antibody specifically detects GTF2IRD2 in Human samples. It is validated for Flow Cytometry
Specifications
| GTF2IRD2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 84163 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| PCRP-GTF2IRD2-1B12 | |
| Flow Cytometry | |
| FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha | |
| Recombinant full-length human GTF2IRD2 protein (Uniprot: Q86UP8) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction