missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2IRD2 Antibody (PCRP-GTF2IRD2-1B12) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
GTF2IRD2 Monoclonal specifically detects GTF2IRD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
Specifications
Specifications
| Antigen | GTF2IRD2 |
| Applications | Western Blot, Flow Cytometry, Peptide Array |
| Classification | Monoclonal |
| Clone | PCRP-GTF2IRD2-1B12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1-2 ug/mL, Flow Cytometry : 1-2 ug/million cells, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?