missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GTF2IRD1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16376994
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16376994 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16376994 Supplier Invitrogen™ Supplier No. PA578295

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: HeLa, HeLa nuclear Predicted Reactivity: Mouse (85%), Rat (85%), Rhesus Monkey (99%), Bovine (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Williams-Beuren syndrome (WBS) is a developmental disorder caused by the hemizygous microdeletion on chromosome 7q11.23. WBS is an autosomal dominant genetic condition that is characterized by physical, cognitive and behavioral traits. The physical traits associated with WBS include facial dysmorphology, vascular stenoses, growth deficiencies, dental anomalies and neurologic and musculoskeletal abnormalities. Mild retardation, a weakness in visual-spatial skills, anxiety and a short attention span are typical cognitive and behavioral traits of WBS patients. The WBSCR11 gene is located within the WBS deletion and may contribute to the developmental symptoms found in WBS because of a loss of the encoded transcription factor. WBSCR11 is also designated GRF2IRD1, GTF3, Cream1 and MusTRD1 in human and BEN in mouse, due to slight differences in gene structure. WBSCR11 is expressed in all adult tissues as several variants and has discrete spatial and temporal expression during embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2IRD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene GTF2IRD1
Gene Accession No. Q9UHL9
Gene Alias 1700012P16Rik; Alb/c-myc line 166.8; Alb-c-myc line 166.8; BEN; Binding factor for early enhancer; c-myc line 166.8; CREAM1; ESTM9; general transcription factor 3; general transcription factor II I repeat domain-containing 1; General transcription factor III; general transcription factor II-I repeat domain-containing protein 1; GTF2I repeat domain containing 1; GTF2I repeat domain-containing 1; GTF2I repeat domain-containing protein 1; Gtf2il; Gtf2ird1; GTF3; hMusTRD1alpha1; Muscle TFII-I repeat domain-containing protein 1; muscle TFII-I repeat domain-containing protein 1 alpha 1; MUSTRD1; MusTRD1/BEN; RBAP2; slow-muscle-fiber enhancer-binding protein; Tg(Alb1-Myc)166.8Sst; transcription factor GTF3 alpha 2; transcription factor GTF3 gamma 2; USE B1-binding protein; WBS; WBSCR11; WBSCR12; Williams-Beuren syndrome chromosomal region 11 protein; Williams-Beuren syndrome chromosomal region 12 protein; Williams-Beuren syndrome chromosome region 11; X83320
Gene Symbols GTF2IRD1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GTF2IRD1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9569
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.