missing translation for 'onlineSavingsMsg'
Learn More

GTF2IRD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-38300-20ul

 View more versions of this product

Product Code. 30229771

  • £145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GTF2IRD1 Polyclonal antibody specifically detects GTF2IRD1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GTF2IRD1
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
BEN, CREAM1, General transcription factor III, general transcription factor II-I repeat domain-containing protein 1, GTF2I repeat domain containing 1, GTF2I repeat domain-containing protein 1, GTF3GTF2I repeat domain-containing 1, hMusTRD1alpha1, Muscle TFII-I repeat domain-containing protein 1, muscle TFII-I repeat domain-containing protein 1 alpha 1, MusTRD1, MusTRD1/BEN, MUSTRD1general transcription factor 3, RBAP2WBSCR12binding factor for early enhancer, Slow-muscle-fiber enhancer-binding protein, USE B1-binding protein, WBS, WBSCR11, Williams-Beuren syndrome chromosomal region 11 protein, Williams-Beuren syndrome chromosomal region 12 protein, Williams-Beuren syndrome chromosome region 11
Recombinant fusion protein containing a sequence corresponding to amino acids 660-959 of human GTF2IRD1 (NP_057412.1).,, Sequence:, SLGFSPPALPPERDSGDPLVDESLKRQGFQENYDARLSRIDIANTLREQVQDLFNKKYGEALGIKYPVQVPYKRIKSNPGSVIIEGLPPGIPFRKPCTFGSQNLERILAVADKIKFTVTRPFQGLIPKPDEDDANRLGEKVILREQVKELFNEKYGEALGLNRPVLVPYKLIRDSPDAVEVTGLPDDIPFRNPNTYDIHRLEKILKAREHVRMVIINQLQPFAEICNDAKVPAKDSSIPKRKRKRVSEGNSVSSSSSSSSSSSSNPDSVASANQISLVQWPMYMVDYAGLNVQLPGPLNY
20 μL
Primary
Human, Mouse, Rat
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
9569
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.