Learn More
Invitrogen™ GTF2I Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117110
Description
Positive Samples: BT-474, Mouse brain Immunogen sequence: FESNPEFLYV EGLPEGIPFR SPTWFGIPRL ERIVRGSNKI KFVVKKPELV ISYLPPGMAS KINTKALQSP KRPRSPGSNS KVPEIEVTVE GPNNNNPQTS AVRTPTQTNG SNVPFKPRGR EFSFEAWNAK ITDLKQKVEN LFNEKCGEAL GLKQAVKVPF ALFESFPEDF YVEGLPEGVP FRRPSTFGIP RLEKILRNKA KIKFIIKKPE M.
This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
Specifications
| GTF2I | |
| Polyclonal | |
| Unconjugated | |
| GTF2I | |
| 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14886, 2969, 353256 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78347, Q5U2Y1, Q9ESZ8 | |
| GTF2I | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-810 of human GTF2I (NP_0011571081). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.