missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2H4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540448
This item is not returnable.
View return policy
Description
Peptide sequence: ALWVKKEFSK AQEESTGLLS GLRIWHTQLL PGGLQGLILN PIFRQNLRIA Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.
GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Specifications
| GTF2H4 | |
| Polyclonal | |
| Unconjugated | |
| GTF2H4 | |
| AW545633; basic transcription factor 2 52 kDa subunit; BTF2 p52; general transcription factor II H, polypeptide 4; general transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; GTF2H4; p52; TFB2; TFIIH; TFIIH basal transcription factor complex p52 subunit; TFIIH transcription/DNA repair factor p52 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14885, 2968 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O70422, Q92759 | |
| GTF2H4 | |
| Synthetic peptide directed towards the N-terminal of human GTF2H4 (aa 72-121). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction