missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19869-1-AP-20UL
This item is not returnable.
View return policy
Description
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.Specifications
| GTF2H3 | |
| Polyclonal | |
| Unconjugated | |
| GTF2H3 | |
| BTF2, BTF2 p34, GTF2H3, TFB4, TFIIH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2967 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13889 | |
| GTF2H3 | |
| GTF2H3 Fusion Protein Ag7131 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction