missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H2D Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £359.00
Specifications
| Antigen | GTF2H2D |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637388
|
Novus Biologicals
NBP3-05592-100ul |
100 μg |
£359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612068
|
Novus Biologicals
NBP3-05592-20ul |
20 μg |
£149.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF2H2D Polyclonal antibody specifically detects GTF2H2D in Rat samples. It is validated for Western BlotSpecifications
| GTF2H2D | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| DKFZp686P18101, FLJ77682, General transcription factor IIH polypeptide 2-like protein, general transcription factor IIH subunit 2-like protein, general transcription factor IIH, polypeptide 2C, GTF2H2D, MGC102806, MGC198575 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human GTF2H2D (NP_001035955.1). MDEEPERTKRWEGGYERTWEILKEDESGSLKATIEDILFKAKRKRVFEHHGQVRLGMMRHLYVVVDGSRTMEDQDLKPNRLTCTLKLLEYFVEEYFDQNPISQIGIIVTKSKRAEKLTELSGNPRKHITSLKEAVDMTCHGEPSLYNSLSMAMQTLKHMPGHTSREVLIIFSSLTTCDPSNIYDLIKTLKAAKIRVSVIGLSAEVRVCTVLARETGGTYHVILDESHYKELLTHHLSPPPASSSSECSLIRMGFPQHTIA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 730394 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title