missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2H2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548199
This item is not returnable.
View return policy
Description
GTF2H2C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Specifications
| GTF2H2C | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6P1K8 | |
| GTF2H2C | |
| KLH conjugated synthetic peptide between 314-343 amino acids from the C-terminal region of human GTF2H2C. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| GTF2H2C | |
| general transcription factor IIH polypeptide 2-like protein; General transcription factor IIH subunit 2-like protein; general transcription factor IIH, polypeptide 2C; GTF2H2 family member C; GTF2H2C; GTF2H2C_2; GTF2H2D | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 728340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction