missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H2C Antibody (PCRP-GTF2H2C-2C9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08896R
This item is not returnable.
View return policy
Description
GTF2H2C Monoclonal antibody specifically detects GTF2H2C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| GTF2H2C | |
| Monoclonal | |
| DyLight 550 | |
| General transcription factor IIH polypeptide 2-like protein, general transcription factor IIH subunit 2-like protein, general transcription factor IIH, polypeptide 2C, GTF2H2C_2 | |
| Recombinant full-length human GTF2H2C protein (Uniprot: Q6P1K8) | |
| 100 μg | |
| Direct Reversal of DNA Damage | |
| 728340 | |
| Store at 4°C in the dark. | |
| IgG2c |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-GTF2H2C-2C9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction